LIF372153589 Life Chemicals Inc.

N-(4-chloro-6-methylpyrimidin-2-yl)acetamide

  • MDL number MFCD00461610
  • Cas Number : 89694-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart