LIF372153595 Life Chemicals Inc.

[3-(3-pyridin-3-yl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]amine

  • MDL number MFCD16652761
  • Cas Number : 1204296-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart