LIF372153603 Life Chemicals Inc.

3-(4-methoxyphenyl)pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD16652766
  • Cas Number : 1204296-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart