LIF372153606 Life Chemicals Inc.

4-thioxo-1,3,4,5,6,7-hexahydro-2H-cyclopenta[d]pyrimidin-2-one

  • MDL number MFCD12917535
  • Cas Number : 21582-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart