LIF372153643 Life Chemicals Inc.

(E)-2-(4-methylphenyl)ethylenesulfonyl chloride

  • MDL number MFCD04973464
  • Cas Number : 98821-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart