LIF372153648 Life Chemicals Inc.

6-chloro-2-methylpyrimidin-4-amine

  • MDL number MFCD07782029
  • Cas Number : 1749-68-4
  • Molecular Weight : 143.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart