LIF372153661 Life Chemicals Inc.

2-(3-methyl-6-oxo-1,6-dihydropyridazin-1-yl)acetic acid

  • MDL number MFCD07186386
  • Cas Number : 412018-60-1
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart