LIF372153666 Life Chemicals Inc.

1-(4-chlorophenyl)pyrimidine-2,4,6(1H,3H,5H)-trione

  • MDL number MFCD00447898
  • Cas Number : 90767-54-7
  • Molecular Weight : 238.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart