LIF372153689 Life Chemicals Inc.

ethyl 5-phenyl-1,3-oxazole-2-carboxylate

  • MDL number MFCD11975682
  • Cas Number : 13575-16-1
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart