LIF372153692 Life Chemicals Inc.

ethyl 5-(4-methoxyphenyl)-1,3-oxazole-2-carboxylate

  • MDL number MFCD21091666
  • Cas Number : 1441-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart