LIF372153703 Life Chemicals Inc.

4-[3-(4-methoxyphenyl)-6-oxopyridazin-1(6H)-yl]butanoic acid

  • MDL number MFCD21091674
  • Cas Number : 1283109-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart