LIF372153720 Life Chemicals Inc.

6-(4-methoxyphenyl)-3-methylimidazo[2,1-b][1,3]thiazole-2-carboxylic acid

  • MDL number MFCD11221793
  • Cas Number : 1131580-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart