LIF372153722 Life Chemicals Inc.

1-[6-(2-methoxyphenyl)pyridazin-3-yl]piperidine-4-carboxylic acid

  • MDL number MFCD22587495
  • Cas Number : 1286697-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart