LIF372153723 Life Chemicals Inc.

3-methyl-6-(2-piperazin-1-ylethoxy)pyridazine

  • MDL number MFCD21091684
  • Cas Number : 1286713-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart