LIF372153724 Life Chemicals Inc.

2-(2-piperazin-1-ylethyl)pyridazin-3(2H)-one

  • MDL number MFCD17233976
  • Cas Number : 1341994-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart