LIF372153745 Life Chemicals Inc.

4-ethoxy-3-methylbenzenesulfonyl chloride

  • MDL number MFCD08752805
  • Cas Number : 69129-64-2
  • Molecular Weight : 234.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart