LIF372153747 Life Chemicals Inc.

6-morpholin-4-ylpyridazine-3-carboxylic acid

  • MDL number MFCD08235211
  • Cas Number : 914637-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart