LIF372153752 Life Chemicals Inc.

2,6-dioxo-1-phenyl-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid

  • MDL number MFCD03232461
  • Cas Number : 723-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart