LIF372153829 Life Chemicals Inc.

(4a,6a-Dimethyl-tetrahydro-1,4-dioxa-6b-aza-cyclopenta[cd]pentalen-2a-yl)- methanol

  • MDL number MFCD00054537
  • Cas Number : 60204-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart