LIF372153830 Life Chemicals Inc.

[1-(4-methoxyphenyl)-2-phenylethyl]methylamine

  • MDL number MFCD00025823
  • Cas Number : 93027-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart