LIF372153835 Life Chemicals Inc.

5-Methyl-4,5,6,7-tetrahydro-2,1-benzisoxazol-3-amine

  • MDL number MFCD08143670
  • Cas Number : 879644-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart