LIF372153844 Life Chemicals Inc.

3-(4-chloro-3,5-dimethyl-1H-pyrazol-1-yl)butanoic acid

  • MDL number MFCD05668685
  • Cas Number : 956354-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart