LIF372153851 Life Chemicals Inc.

3,5-Dimethyl-4-(pyrrolidin-1-ylsulfonyl)-1H-pyrazole

  • MDL number MFCD06496381
  • Cas Number : 890092-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart