LIF372153863 Life Chemicals Inc.

6-Chloro-N-(4-chloro-2-methylphenyl)-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine

  • MDL number MFCD11518858
  • Cas Number : 1040662-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart