LIF372153865 Life Chemicals Inc.

6-Chloro-1-methyl-4-piperidin-1-yl-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD11518860
  • Cas Number : 1040662-63-2
  • Molecular Weight : 251.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart