LIF372153868 Life Chemicals Inc.

6-Chloro-N-(3-chloro-4-methylphenyl)-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine

  • MDL number MFCD11518863
  • Cas Number : 1040662-70-1
  • Molecular Weight : 308.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart