LIF372153873 Life Chemicals Inc.

6-Chloro-N-(3-chlorophenyl)-1-phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine

  • MDL number MFCD11518867
  • Cas Number : 1040662-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart