LIF372153879 Life Chemicals Inc.

(5-Bromo-2-methylamino-phenyl)-phenyl-methanone

  • MDL number MFCD06326206
  • Cas Number : 39573-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart