LIF372153882 Life Chemicals Inc.

(2-amino-5-chlorophenyl)(4-fluorophenyl)methanone

  • MDL number MFCD07780289
  • Cas Number : 784-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart