LIF372153883 Life Chemicals Inc.

3-chloro-1H-pyrazolo[3,4-b]pyridine-5-carbaldehyde

  • MDL number MFCD22587539
  • Cas Number : 1351613-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart