LIF372153890 Life Chemicals Inc.

6-Amino-5-(2-furoyl)-1,3-dimethylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD07803525
  • Cas Number : 890093-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart