LIF372153910 Life Chemicals Inc.

3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine

  • MDL number MFCD09863958
  • Cas Number : 230305-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart