LIF372153927 Life Chemicals Inc.

(7-Methoxy-2-methyl-1H-indol-3-yl)-acetic acid

  • MDL number MFCD09863997
  • Cas Number : 50995-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart