LIF372153949 Life Chemicals Inc.

1-[4-Cyano-2-(2-thienyl)-1,3-oxazol-5-yl]piperidine-4-carboxylic acid

  • MDL number MFCD11518871
  • Cas Number : 1105195-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart