LIF372153953 Life Chemicals Inc.

6-Chloro-4-oxo-4H-chromene-3-carbaldehyde

  • MDL number MFCD00139138
  • Cas Number : 42248-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart