LIF372153959 Life Chemicals Inc.

5-(4-Aminophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD06496340
  • Cas Number : 889302-74-3
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart