LIF372153979 Life Chemicals Inc.

9-Methyl-9H-purin-6-ol

  • MDL number MFCD00127899
  • Cas Number : 875-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart