LIF372153997 Life Chemicals Inc.

2-Chloro-9-methyl-9H-purine

  • MDL number MFCD07440150
  • Cas Number : 2346-73-8
  • Molecular Weight : 168.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart