LIF372153999 Life Chemicals Inc.

1-(5-Amino-2-chloro-6-methylpyrimidin-4-yl)piperidine-4-carboxylic acid

  • MDL number MFCD07440194
  • Cas Number : 890094-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart