LIF372154002 Life Chemicals Inc.

(2-Chloro-5-nitro-pyrimidin-4-yl)-methyl-amine

  • MDL number MFCD07440210
  • Cas Number : 89283-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart