LIF372154003 Life Chemicals Inc.

(2-Chloro-5-nitro-pyrimidin-4-yl)-cyclopentyl-amine

  • MDL number MFCD07440225
  • Cas Number : 330550-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart