LIF372154017 Life Chemicals Inc.

5-Hydroxy-2-oxo-1,2,5,6,7,8-hexahydroquinoline-3-carboxylic acid

  • MDL number MFCD08692071
  • Cas Number : 923219-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart