LIF372154025 Life Chemicals Inc.

2-Chloro-5,6,7,8-tetrahydroquinolin-5-ol

  • MDL number MFCD08692080
  • Cas Number : 501917-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart