LIF372154033 Life Chemicals Inc.

methyl 4-(2-aminothiazol-4-yl)benzoate

  • MDL number MFCD03030162
  • Cas Number : 206555-77-3
  • Molecular Weight : 234.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart