LIF372154059 Life Chemicals Inc.

2-[(Chloroacetyl)amino]benzoic acid

  • MDL number MFCD00029924
  • Cas Number : 14422-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart