LIF372154065 Life Chemicals Inc.

1-methanesulfonylpyrrolidine-2-carboxylic acid

  • MDL number MFCD06347033
  • Cas Number : 360045-22-3
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart