LIF372154089 Life Chemicals Inc.

2,5-Dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD01830401
  • Cas Number : 2199-63-5
  • Molecular Weight : 123.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart