LIF372154109 Life Chemicals Inc.

2-Trifluoromethyl-pyrimidin-4-ylamine

  • MDL number MFCD08436606
  • Cas Number : 672-42-4
  • Molecular Weight : 163.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart