LIF372154113 Life Chemicals Inc.

4-Amino-2-trifluoromethyl-pyrimidine-5-carboxylic acid

  • MDL number MFCD08436634
  • Cas Number : 777-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart