LIF372154116 Life Chemicals Inc.

4-Chloro-6-trifluoromethyl-pyrimidine

  • MDL number MFCD04972771
  • Cas Number : 37552-81-1
  • Molecular Weight : 182.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart